missing translation for 'onlineSavingsMsg'
Learn More

Methylene Blue Milk Smear Stain, Ricca Chemical

Catalog No. 489016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
489016 500 mL Amber Glass Bottle
489032 1 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 489016 Supplier Ricca Chemical Company Supplier No. 489016

Modified Newman-Lampert Formulation, for Microscopic Bacterial Examination of Milk

Specifications

Chemical Name or Material Methylene Blue Milk Smear Stain
Name Note Modified Newman-Lampert Formulation
CAS 79-34-5, 64-17-5, 64-19-7, 7732-18-5, 65-56-1, 7220-79-3
CAS Min % 59.0, 33.18, 3.53, 2.18, 0.5
CAS Max % 61.0, 34.53, 3.67, 2.27, 0.52
Grade Laboratory
Packaging Amber Glass Bottle
Color Dark Blue
Quantity 500 mL
Physical Form Liquid