missing translation for 'onlineSavingsMsg'
Learn More

Methyl decanoate, EMD-Millipore

Fatty acid methyl esters

Supplier:  Technidata 1096370005

Numéro de catalogue. M1096370005


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Methyl decanoate
110-42-9
Density: 0.87g/cm3 (20°)
C11H22O2
Methyl caprate, Capric acid methyl ester, Decanoic acid methyl ester
CCCCCCCCCC(=O)OC
186.30
8050
Glass vials
0.7 %(V)
≥99.5%
MFCD00009580
YRHYCMZPEVDGFQ-UHFFFAOYSA-N
methyl decanoate
5 mL
186.29

Identifiants chimiques

110-42-9
186.30
YRHYCMZPEVDGFQ-UHFFFAOYSA-N
8050
CCCCCCCCCC(=O)OC
C11H22O2
MFCD00009580
Methyl caprate, Capric acid methyl ester, Decanoic acid methyl ester
methyl decanoate
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

Recommended Storage : Store at 15° to 25°C

FDS
Documents

Documents

Promotions

Promotions