missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl α-D-Mannopyranoside, 99.3%, MP Biomedicals
Supplier: MP Biomedicals Inc 0215393710
Spécifications
| Methyl α-D-Mannopyranoside | |
| White | |
| C7H14O6 | |
| 10 g | |
| HOVAGTYPODGVJG-UHFFFAOYNA-N | |
| 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
| 101798 | |
| 194.2 |
| 617-04-9 | |
| ∽ 99% | |
| MFCD00063262 | |
| methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
| COC1OC(CO)C(O)C(O)C1O | |
| 194.18 | |
| CHEBI:43943 | |
| Crystalline Powder |
Identifiants chimiques
| 617-04-9 | |
| 194.18 | |
| HOVAGTYPODGVJG-UHFFFAOYNA-N | |
| 101798 | |
| 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
| C7H14O6 | |
| MFCD00063262 | |
| methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
| CHEBI:43943 | |
| COC1OC(CO)C(O)C(O)C1O |
Sécurité et manipulation
Recommended Storage : Store at room temperature.