missing translation for 'onlineSavingsMsg'
Learn More

Methyl α-D-Mannopyranoside, 99.3%, MP Biomedicals

Catalog No. p-4405023
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15393710 10 g
ICN15393725 25 g
ICN15393780 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15393710 Supplier MP Biomedicals Inc Supplier No. 0215393710
Only null left

Chemical Identifiers

CAS 617-04-9
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00063262
InChI Key HOVAGTYPODGVJG-UHFFFAOYNA-N
Synonym methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside
PubChem CID 101798
ChEBI CHEBI:43943
IUPAC Name 2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1OC(CO)C(O)C(O)C1O

Specifications

Color White
Assay Percent Range ∽ 99%
Quantity 10 g
Formula Weight 194.2
Physical Form Crystalline Powder
Chemical Name or Material Methyl α-D-Mannopyranoside
Recommended Storage Store at room temperature.