missing translation for 'onlineSavingsMsg'
Learn More

Methyl 4-bromobenzoate, 98+%

CAS: 619-42-1 | C8H7BrO2 | 215.046 g/mol

Supplier:  Thermo Scientific Chemicals A1551714

 Afficher plus de versions de ce produit

Numéro de catalogue. AAA1551714


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Methyl 4-bromobenzoate is used as a starting material in the synthesis of three-carbon-bridged 5-substituted furo[2,3-d]pyrimidine and 6-substituted pyrrolo[2,3-d]pyrimidine and methyl 4-tri-n-butylstannylbenzoate. It is also used to stimulate microbial dechlorination of polychlorinated biphenyls. Further, it is used in the preparation of stable radioiodinating reagent to label monoclonal antibodies for radiotherapy of cancer.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Spécifications

Spécifications

Methyl 4-bromobenzoate
77°C to 81°C
C8H7BrO2
25 g
14,1408
Insoluble in water.
COC(=O)C1=CC=C(C=C1)Br
215.046
215.05
619-42-1
1.69
MFCD00013531
2045132
benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate
CZNGTXVOZOWWKM-UHFFFAOYSA-N
methyl 4-bromobenzoate
12081
≥98%

Identifiants chimiques

619-42-1
215.046
CZNGTXVOZOWWKM-UHFFFAOYSA-N
12081
COC(=O)C1=CC=C(C=C1)Br
C8H7BrO2
MFCD00013531
benzoic acid, 4-bromo-, methyl ester, methyl p-bromobenzoate, 4-bromobenzoic acid methyl ester, p-bromobenzoic acid, methyl ester, unii-ub1ysj5xar, ub1ysj5xar, benzoic acid, p-bromo-, methyl ester, pubchem3962, methyl-4-bromobenzoate, methyl p-bromo benzoate
methyl 4-bromobenzoate
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 210-596-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only