missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-fluorobenzoate, 98%

CAS: 394-35-4 | C8H7FO2 | 154.14 g/mol

$26.48 - $88.10

Identifiants chimiques

CAS 394-35-4
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00017913
InChI Key QAFJIJWLEBLXHH-UHFFFAOYSA-N
Synonym methyl o-fluorobenzoate, methyl-2-fluorobenzoate, 2-fluorobenzoic acid methyl ester, benzoic acid, 2-fluoro-, methyl ester, o-fluorobenzoic acid, methyl ester, benzoic acid, o-fluoro-, methyl ester, 2-fluoro-benzoic acid methyl ester, methyl2-fluorobenzoate, pubchem3565, acmc-209j5e
PubChem CID 67854
IUPAC Name methyl 2-fluorobenzoate
SMILES COC(=O)C1=CC=CC=C1F
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantité  
AAA1613306
Afficher les documents
Thermo Scientific Chemicals
A1613306
5 g
chaque for $26.48
Il en reste null
 
AAA1613314
Afficher les documents
Thermo Scientific Chemicals
A1613314
25 g
chaque for $88.10
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

394-35-4
154.14
QAFJIJWLEBLXHH-UHFFFAOYSA-N
67854
COC(=O)C1=CC=CC=C1F
C8H7FO2
MFCD00017913
methyl o-fluorobenzoate, methyl-2-fluorobenzoate, 2-fluorobenzoic acid methyl ester, benzoic acid, 2-fluoro-, methyl ester, o-fluorobenzoic acid, methyl ester, benzoic acid, o-fluoro-, methyl ester, 2-fluoro-benzoic acid methyl ester, methyl2-fluorobenzoate, pubchem3565, acmc-209j5e
methyl 2-fluorobenzoate

Spécifications

394-35-4
109°C to 110°C (35 mmHg)
Aromatic
1.503
5 g
methyl o-fluorobenzoate, methyl-2-fluorobenzoate, 2-fluorobenzoic acid methyl ester, benzoic acid, 2-fluoro-, methyl ester, o-fluorobenzoic acid, methyl ester, benzoic acid, o-fluoro-, methyl ester, 2-fluoro-benzoic acid methyl ester, methyl2-fluorobenzoate, pubchem3565, acmc-209j5e
COC(=O)C1=CC=CC=C1F
154.14
154.14
Methyl 2-fluorobenzoate
1.21
93°C (199°F)
C8H7FO2
MFCD00017913
1862493
QAFJIJWLEBLXHH-UHFFFAOYSA-N
methyl 2-fluorobenzoate
67854
98%
Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement
H315-H319-H227
Causes skin irritation.
Causes serious eye irritation.
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

missing translation for 'einecsNumber' : 206-894-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

FDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only