missing translation for 'onlineSavingsMsg'
Learn More

Methanol Anhydrous ACS, MilliporeSigma™

Supplier:  Technidata MX04873

 Afficher plus de versions de ce produit

Numéro de catalogue. MMX04873


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Methanol
67-56-1,7732-18-5
Metal Can
CH3OH
0.03% max
OKKJLVBELUTLKV-UHFFFAOYSA-N
methanol
887
32.04
Anhydrous
20 L
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CO
32.04
CHEBI:17790
ACS Reagent

Identifiants chimiques

67-56-1
32.04
OKKJLVBELUTLKV-UHFFFAOYSA-N
887
methanol
CH4O
MFCD00004595
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
CHEBI:17790
CO
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions