Learn More
Promega Maxwell™ CSC RNA Blood Kit DFS Item
The Maxwell CSC RNA Blood Kit is intended for use as an IVD medical device, in combination with the Maxwell CSC Instrument, to provide an easy method for efficient, automated purification of RNA from 2.5mL fresh human whole blood collected in EDTA tubes.
Supplier:  Promega AS1410
Numéro de catalogue. PRAS1410
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Automated RNA Isolation from Blood for In Vitro Diagnostic Assays
  • Purifies high-quality RNA from 1-16 whole blood samples
  • Intended for in vitro diagnostic use in combination with the Maxwell(R) CSC Instrument
  • High-quality RNA purification method that is manufactured under cGMP
Spécifications
Spécifications
15°C to 30°C
48 Preps
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS