Learn More
MAPA™ TempIce 700 Nitrile-Coated Knitted Thermal Protection Gloves
Nitrile coating combined with an insulated liner provides thermal protection, while also remaining comfortable and flexible in temperatures down to -10°C.
Supplier:  MAPA™ 34700018
 Afficher plus de versions de ce produit
Numéro de catalogue. 19-046-561
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Nitrile coating provides non-slip grip capabilites in wet or dry conditions
  • Insulated liner protects hands in temperatures down to -10°C
  • Materials remain flexible and comfortable, even in low temperatures
  • Silicone free to prevent traces and defects on metal sheets or glass
  • Washable for up to 5 cleaning cycles
Spécifications
Spécifications
Nitrile
No Texture
8
Blue
TempIce 700
-10°C
25 cm
Black
Specific
Construction, Maintenance, Order Preparation, and Waste Collection
Knit
14°F
9.8 in.
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS