missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ Blue-Grip™ LL-301 Heavyweight Latex Gloves, Interlock Lining

Heavy gauge, 45 mil thick rubber latex gloves provide mechanical and chemical protection, high cut resistance and tear strength.

Supplier:  MAPA™ 301427

 Afficher plus de versions de ce produit

Numéro de catalogue. 11-392-40B



Il en reste null
Découvrez plus d'offres spéciales

Avertissement:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Description

Description

Rated excellent for handling acids, “mild” caustics, alcohols, sanitizing agents and detergents, as well as other chemical, mechanical and thermal hazards. Ideal for handling frozen food, hot containers, sharp edges, abrasives, cutting and tearing hazards, sheet metal and plastic resins.

  • Heavy-duty, liquid-proof gloves with a double-dipped nonabrasive, nonslip grip
  • Interlock-knit cotton lining absorbs perspiration
  • For intermittent exposure from -17° to +100°C (0° to 212°F)
  • Gloves feature a natural rubber latex coating firmly bonded to a reinforcing, 9 oz. (255g) cotton knit lining
  • Hand contoured with no seams in the work area
  • Light blue with a medium blue grip
  • Launderable

Spécifications

Spécifications

21 CFR 170-199
No
Medium
1.14 mm
Pinked
Interlock knit
301427
30.5 cm
Blue
Latex
45 mil
Handling acids, “mild” caustics, alcohols, sanitizing agents and detergents, as well as other chemical, mechanical and thermal hazards, handling frozen food, hot containers, sharp edges, abrasives, cutting and tearing hazards, sheet metal and Pl
Etched
Lined
12 in.
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions