missing translation for 'onlineSavingsMsg'
Learn More

MAPA™ Blue-Grip™ LL-301 Heavyweight Latex Gloves, Interlock Lining

Heavy gauge, 45 mil thick rubber latex gloves provide mechanical and chemical protection, high cut resistance and tear strength.

$7.98 - $9.04

Specifications

Certifications/Compliance 21 CFR 170-199
Color Blue
Latex Free No
Material Latex
Thickness (English) 45 mil
View More Specs

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Products 2
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
11-392-40A
View Documents
MAPA™
301426
Small
Pair for $7.98
Case of 48 Pair for $98.88
Only null left
 
11-392-40B
View Documents
MAPA™
301427
Medium
Pair for $9.04
Case of 48 Pair for $95.04
Only null left
 
Description

Description

Rated excellent for handling acids, “mild” caustics, alcohols, sanitizing agents and detergents, as well as other chemical, mechanical and thermal hazards. Ideal for handling frozen food, hot containers, sharp edges, abrasives, cutting and tearing hazards, sheet metal and plastic resins.

  • Heavy-duty, liquid-proof gloves with a double-dipped nonabrasive, nonslip grip
  • Interlock-knit cotton lining absorbs perspiration
  • For intermittent exposure from -17° to +100°C (0° to 212°F)
  • Gloves feature a natural rubber latex coating firmly bonded to a reinforcing, 9 oz. (255g) cotton knit lining
  • Hand contoured with no seams in the work area
  • Light blue with a medium blue grip
  • Launderable

Specifications

Specifications

21 CFR 170-199
No
45 mil
Handling acids, “mild” caustics, alcohols, sanitizing agents and detergents, as well as other chemical, mechanical and thermal hazards, handling frozen food, hot containers, sharp edges, abrasives, cutting and tearing hazards, sheet metal and Pl
Etched
Lined
30.5 cm
Blue
Latex
1.14 mm
Pinked
Interlock knit
12 in.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'