missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MAbPac™ Reversed Phase HPLC Columns
GREENER_CHOICE

Numéro de catalogue. 088648
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
Stationary Phase:
Phenyl (Ph)
PSDVB (Polystyrene Divinylbenzene)
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric) Diameter (Metric) Stationary Phase
088648 50 mm 2.1 mm Phenyl (Ph)
088644 100 mm 3 mm Phenyl (Ph)
088645 50 mm 3 mm Phenyl (Ph)
088646 10 mm 3 mm Phenyl (Ph)
088647 100 mm 2.1 mm Phenyl (Ph)
088649 10 mm 2.1 mm Phenyl (Ph)
03251884 50 mm 1 mm PSDVB (Polystyrene Divinylbenzene)
03251885 100 mm 1 mm PSDVB (Polystyrene Divinylbenzene)
03251886 150 mm 1 mm PSDVB (Polystyrene Divinylbenzene)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 088648 Supplier Thermo Scientific™ Supplier No. 088648
Only null left

Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.

Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.

Compatible with organic solvent used in HPLC and MS under RP conditions
  • Resin is based on a hydrophobic supermacroporous 4μm polymer particles
  • Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)
Efficient with low carry over
  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over
Designed for reversed-phase biomolecule LC and LC/MS separations
  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Max. Pressure 4000 psi (275 bar)
Particle Size 4 μm
pH 0 to 14
Pore Size 1500 Å
Quantity Each
Temperature <110°C
Stationary Phase Phenyl (Ph)
Product Line MAbPac
Packing Material DVB (Divinylbenzene) Polymer
Column Type Reversed Phase
Phase Reversed Phase
Technique Bio-LC
Green Features FSC
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.