missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lutetium(III) acetate hydrate, REacton™, 99.9% (REO)
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Physical Form | Crystalline Aggregates |
| Quantity | 1 g |
| Solubility Information | Soluble in water. |
| Formula Weight | 352.11 (Anhydrous) |
| Percent Purity | 99.9% |
| Grade | Reacton |
| Sensitivity | Hygroscopic |
| Assay | (REO) |
| Chemical Name or Material | Lutetium(III) acetate hydrate |
Sécurité et manipulation
| EINECSNumber | 242-566-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only