missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lithium Perchlorate, ACS Reagent, ≥95.0%, Honeywell™ Fluka™
ACS Reagent,≥95.0%
Supplier: Honeywell-Fluka 205281100G
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécifications
| Lithium Perchlorate | |
| 3-9-7791 | |
| 6.0 to 9.5 (at 106.4g/L 25°C) | |
| ClH6LiO7 | |
| MFCD00011079 | |
| lithium perchlorate, perchloric acid, lithium salt, lithiumperchlorate, unii-q86se98c9c, lithium 1+ ion perchlorate, lithium perchlorate, anhydrous, lithium cloricum, lithium per chlorate, liclo4, perchloric acid, lithium salt 1:1 | |
| [Li+].[O-]Cl(=O)(=O)=O | |
| 160.43 | |
| 106.39g/mol | |
| ACS Reagent | |
| Granules |
| 236°C | |
| White | |
| 100 g | |
| LiClO4 | |
| UN1481 | |
| JAWYRNYHJJDXHX-UHFFFAOYSA-M | |
| lithium;perchlorate | |
| 23665649 | |
| ≥95.0% | |
| Poly Bottle | |
| 2.420g/cu.cm |
Identifiants chimiques
| 3-9-7791 | |
| 160.43 | |
| JAWYRNYHJJDXHX-UHFFFAOYSA-M |
| ClH6LiO7 | |
| MFCD00011079 |
Sécurité et manipulation
P220-P261-P305 + P351 + P338
H272-H315-H319-H335
missing translation for 'dotInformation' : 5.1/PGII
missing translation for 'einecsNumber' : 232-237-2