missing translation for 'onlineSavingsMsg'
Learn More

Lithium Perchlorate, ACS Reagent, ≥95.0%, Honeywell™ Fluka™

Catalog No. p-7120524
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
Packaging:
Poly Bottle
Catalog No. Quantity Packaging
60045114 500 g Poly Bottle
1 options

Catalog No. 60045114

Supplier: Honeywell Chemicals 205281500G

Only null left

ACS Reagent,≥95.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 3-9-7791
Molecular Formula ClH6LiO7
Molecular Weight (g/mol) 160.43
MDL Number MFCD00011079
InChI Key JAWYRNYHJJDXHX-UHFFFAOYSA-M

Specifications

Melting Point 236°C
Color White
pH 6.0 to 9.5 (at 106.4g/L 25°C)
Quantity 500 g
Linear Formula LiClO4
UN Number UN1481
Synonym sodium perchlorate monohydrate, sodium perchlorate hydrate, unii-g4h485lhcw, g4h485lhcw, ccris 8893, perchloric acid, sodium salt, monohydrate, sodium hydrate perchlorate, clo4.na.h2o, acmc-1bii1, sodium perchlorate-monohydrate
SMILES [Li+].O.O.O.[O-][Cl](=O)(=O)=O
IUPAC Name sodium;perchlorate;hydrate
PubChem CID 516899
Formula Weight 106.39g/mol
Percent Purity ≥95.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Granules
Density 2.420g/cu.cm
Chemical Name or Material Lithium Perchlorate
Show More Show Less
Hazard Statement P220-P261-P305 + P351 + P338
Precautionary Statement H272-H315-H319-H335
DOTInformation 5.1/PGII
EINECSNumber 232-237-2