missing translation for 'onlineSavingsMsg'
Learn More

Lead Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor L107512KG

 Afficher plus de versions de ce produit

Numéro de catalogue. 18611237


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

10099-74-8
12 kg
ACS
1
99%
Poly Pail

Identifiants chimiques

10099-74-8
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.