missing translation for 'onlineSavingsMsg'
Learn More

Lead Nitrate, Crystal, ACS, 99%, Spectrum™ Chemical

$270.91 - $12265.56

Chemical Identifiers

CAS 10099-74-8
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18611241
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
L107525GM
25 g Amber Glass Bottle
Each for $270.91
Only null left
 
18611236
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
L1075125GM
125 g Poly Bottle
Each for $509.19
Only null left
 
18611238
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
L1075500GM
500 g Poly Bottle
Each for $1,221.00
Only null left
 
18611240
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
L107525KG
2.5 kg Poly Bottle
Each for $3,887.03
Only null left
 
18611237
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
L107512KG
12 kg Poly Pail
Each for $7,434.63
Only null left
 
18611239
missing translation for 'viewDocuments'
Spectrum Chemical Mfg Cor
L107550KGBL
50 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10099-74-8

Specifications

10099-74-8
25 g
ACS
1
99%
Amber Glass Bottle
Videos
SDS
Documents

Documents

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.