Learn More
DWK Life Sciences Kimble™ Kontes™ Flasks with Three Angle Necks and standard taper Joints
Side necks are angled off center neck for greater accessibility
Supplier:  DWK Life Sciences 2955500025
 Afficher plus de versions de ce produit
Numéro de catalogue. K295550-0025
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Spécifications
Spécifications
25 mL
For heating, stirring and solvent stripping without impairing ease of liquid and solid transfer
3
14/20 Center, 14/20 Side
Borosilicate Glass
Glass plug and the patented pressure/vacuum retainer
Kjeldahl
Round
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS