Learn More
DWK Life Sciences Kimble™ ValueWare™ Flat-Bottom Boiling Flasks
Made from Kimax™ Type KG-33 glass.
Supplier: DWK Life Sciences 55055250
Description
- Short neck
- standard taper 24/40 joint
Compliance
Glass meets Type 1, Class A, ASTM™ E-438 standards. Designed to ASTM Specification E1403, Type I, Class 4 specification requirements.
Spécifications
| 3.2 in. | |
| Glass | |
| 13.3 cm | |
| 24/40 | |
| 82 mm |
| 250 mL | |
| 5.23 in. | |
| Short | |
| Flat |