Learn More
DWK Life Sciences Kimble™ Phenolic Caps with Solid PE Liners
Has good resistance to acids, alcohols, alkalis, aqueous solutions, oils, solvents
Supplier: DWK Life Sciences 75202G53400
Avertissement:
Not recommended for hydrocarbon solvents.
Description
- General purpose phenolic screw cap with a solid polyethylene (PE) liner
- Color: black
- Liner material: solid polyethylene (0.040-in.)
- Ideal for use with strong acid or base samples
- Not recommended for hydrocarbon solvents
Spécifications
| Black | |
| 144 Cs. | |
| Phenolic Cap | |
| Polyethylene |
| Phenolic Resin | |
| 53-400 | |
| Bottle |