missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Phenolic Caps with Solid PE Liners

Has good resistance to acids, alcohols, alkalis, aqueous solutions, oils, solvents

$93.26 - $741.39

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Product Type Phenolic Cap
For Use With (Equipment) Bottle
View More Specs

Alert:

Not recommended for hydrocarbon solvents.


Products 14
Catalog Number Mfr. No. Neck Finish Price Quantity  
Catalog Number Mfr. No. Neck Finish Price Quantity  
13-757-137
View Documents
DWK Life Sciences
75202G58400
58-400
Case of 144 Each for $395.39
Only null left
 
13-757-140
View Documents
DWK Life Sciences
75202G89400
89-400
Case of 144 Each for $741.39
Only null left
 
13-757-139
View Documents
DWK Life Sciences
75202G70400
70-400
Case of 144 Each for $544.21
Only null left
 
13-757-130
View Documents
DWK Life Sciences
75202G28400
28-400
Case of 144 Each for $101.33
Only null left
 
13-757-133
View Documents
DWK Life Sciences
75202G43400
43-400
Case of 144 Each for $198.11
Only null left
 
13-757-128
View Documents
DWK Life Sciences
75202G22400
22-400
Case of 144 Each for $141.75
Only null left
 
13-757-129
View Documents
DWK Life Sciences
75202G24400
24-400
Case of 144 Each for $118.31
Only null left
 
13-757-132
View Documents
DWK Life Sciences
75202G38400
38-400
Case of 144 Each for $206.86
Only null left
 
13-757-135
View Documents
DWK Life Sciences
75202G48400
48-400
Case of 144 Each for $305.68
Only null left
 
13-757-131
View Documents
DWK Life Sciences
75202G33400
33-400
Case of 144 Each for $176.68
Only null left
 
13-757-138
View Documents
DWK Life Sciences
75202G63400
63-400
Case of 144 Each for $444.39
Only null left
 
13-757-134
View Documents
DWK Life Sciences
75202G45400
45-400
Case of 144 Each for $300.32
Only null left
 
13-757-136
View Documents
DWK Life Sciences
75202G53400
53-400
Case of 144 Each for $317.00
Only null left
 
13-757-127
View Documents
DWK Life Sciences
75202G20400
20-400
Case of 144 Each for $93.26
Only null left
 
Description

Description

  • Ideal for strong acid or base samples
  • Polyethylene liner
  • General purpose

All caps/liners should be tested to ensure leak and compatibility performance with contents.

Specifications

Specifications

Black
144 Cs.
Bottle
Phenolic Resin
Phenolic Cap
Polyethylene
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'