missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kerosene, Low Odor Deodorized, Reagents
Kerosene, Low Odor Deodorized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc CK190000200E
Spécifications
| Kerosene, Low Odor Deodorized | |
| Colorless to Slight Yellow | |
| Steel Drum | |
| 200 L | |
| Paraffin oil | |
| Liquid |
| 64742-48-9 | |
| 175°C | |
| C12H26-C15H32 | |
| 0.767 | |
| Reagent |
Identifiants chimiques
| 64742-48-9 | |
| C12H26-C15H32 | |
| Paraffin oil |