missing translation for 'onlineSavingsMsg'
Learn More

Kerosene, Low Odor Deodorized, Reagents

Catalog No. p-200021965
Change view
Click to view available options
Quantity:
1 L
4 L
10 L
20 L
200 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01334875 10 L
01334874 1 L
01334873 200 L
01334872 20 L
01334871 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01334875 Supplier Labchem Inc Supplier No. CK19000010E6
Only null left

Kerosene, Low Odor Deodorized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64742-48-9
Molecular Formula C12H26-C15H32
Synonym Paraffin oil

Specifications

Color Colorless to Slight Yellow
Boiling Point 175°C
Packaging Steel Pail
Quantity 10 L
Specific Gravity 0.767
Grade Reagent
Physical Form Liquid
Chemical Name or Material Kerosene, Low Odor Deodorized