missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutylamine Hydrochloride 99.0+%, TCI America™
Supplier: TCI America I0096500G
Spécifications
| Isobutylamine Hydrochloride | |
| 178°C | |
| C4H12ClN | |
| 500 g | |
| BSMNBEHEFWDHJD-UHFFFAOYSA-N | |
| 2-methylpropan-1-amine;hydrochloride | |
| 12206183 | |
| ≥99.0% (T) |
| 5041-09-8 | |
| White | |
| MFCD00060199 | |
| 1-Amino-2-methylpropane Hydrochloride, Isobutylammonium Chloride | |
| CC(C)CN.Cl | |
| 109.60 | |
| 109.60 | |
| Crystalline Powder |
Identifiants chimiques
| 5041-09-8 | |
| 109.60 | |
| BSMNBEHEFWDHJD-UHFFFAOYSA-N | |
| 12206183 | |
| CC(C)CN.Cl |
| C4H12ClN | |
| MFCD00060199 | |
| 1-Amino-2-methylpropane Hydrochloride, Isobutylammonium Chloride | |
| 2-methylpropan-1-amine;hydrochloride |
Sécurité et manipulation
missing translation for 'einecsNumber' : (2)-0132
missing translation for 'tsca' : No