missing translation for 'onlineSavingsMsg'
Learn More

Isobutylamine Hydrochloride 99.0+%, TCI America™

$87.60 - $719.52

Chemical Identifiers

CAS 5041-09-8
Molecular Formula C4H12ClN
Molecular Weight (g/mol) 109.60
MDL Number MFCD00060199
InChI Key BSMNBEHEFWDHJD-UHFFFAOYSA-N
Synonym 1-Amino-2-methylpropane Hydrochloride, Isobutylammonium Chloride
PubChem CID 12206183
IUPAC Name 2-methylpropan-1-amine;hydrochloride
SMILES CC(C)CN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I009625G
View Documents
TCI America
I009625G
25 g N/A
Only null left
 
I0096500G
View Documents
TCI America
I0096500G
500 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5041-09-8
109.60
BSMNBEHEFWDHJD-UHFFFAOYSA-N
12206183
CC(C)CN.Cl
C4H12ClN
MFCD00060199
1-Amino-2-methylpropane Hydrochloride, Isobutylammonium Chloride
2-methylpropan-1-amine;hydrochloride

Specifications

5041-09-8
White
MFCD00060199
1-Amino-2-methylpropane Hydrochloride, Isobutylammonium Chloride
CC(C)CN.Cl
109.60
109.60
Crystalline Powder
178°C
C4H12ClN
25 g
BSMNBEHEFWDHJD-UHFFFAOYSA-N
2-methylpropan-1-amine;hydrochloride
12206183
≥99.0% (T)
Isobutylamine Hydrochloride
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0132

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'