missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron Standard, 200 ppm Fe, Ricca Chemical
$98.00 - $196.94
Chemical Identifiers
| CAS | 10025-77-1 |
|---|---|
| Molecular Formula | Cl3FeH12O6 |
| Molecular Weight (g/mol) | 270.29 |
| MDL Number | MFCD00149712 |
| InChI Key | NQXWGWZJXJUMQB-UHFFFAOYSA-K |
| IUPAC Name | iron(3+) hexahydrate trichloride |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Chemical Identifiers
| 10025-77-1 | |
| 270.29 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
| Cl3FeH12O6 | |
| MFCD00149712 | |
| iron(3+) hexahydrate trichloride |
Specifications
| Natural Poly Bottle | |
| 200ppm Fe | |
| Liquid | |
| MFCD00149712 | |
| 100°C | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
| 270.29 | |
| Iron Standard |
| 10025-77-1 , 7647-01-0 , 7732-18-5 | |
| 1 mL = 0.2 mg Fe | |
| Cl3FeH12O6 | |
| Amber | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| iron(3+) hexahydrate trichloride | |
| 120 mL |