missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iron Standard, 200 ppm Fe, Ricca Chemical
Supplier: Ricca Chemical Company 419016
Specifications
| Iron Standard | |
| 10025-77-1 , 7647-01-0 , 7732-18-5 | |
| 1 mL = 0.2 mg Fe | |
| Cl3FeH12O6 | |
| Amber | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| iron(3+) hexahydrate trichloride | |
| 500 mL |
| Natural Poly Bottle | |
| 200ppm Fe | |
| Liquid | |
| MFCD00149712 | |
| 100°C | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
| 270.29 |
Chemical Identifiers
| 10025-77-1 | |
| 270.29 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
| Cl3FeH12O6 | |
| MFCD00149712 | |
| iron(3+) hexahydrate trichloride |