missing translation for 'onlineSavingsMsg'
Learn More

Iron Standard, 200 ppm Fe, Ricca Chemical

Catalog No. 41904
Change view
Click to view available options
Quantity:
120 mL
500 mL
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
41904 120 mL Natural Poly Bottle
419016 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 41904 Supplier Ricca Chemical Company Supplier No. RI41904

Chemical Identifiers

CAS 10025-77-1 , 7647-01-0 , 7732-18-5
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 200ppm Fe
Equivalence 1 mL = 0.2 mg Fe
Physical Form Liquid
Color Amber
Boiling Point 100°C
Quantity 120 mL
Chemical Name or Material Iron Standard