CAS RN 7647-01-0
Hydrochloric Acid (TraceMetal™ Grade), Fisher Chemical™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution, 1N (Certified), Fisher Chemical™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution, 0.1N in Isopropanol (Certified), Fisher Chemical
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid, Certified ACS Plus, 36.5 to 38.0%, Fisher Chemical™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution, 0.1N (N/10) (Certified), Fisher Chemical™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution, 0.5N (N/2) (Certified), Fisher Chemical™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution, 6N (Certified), Fisher Chemical™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution, 2N (Certified), Fisher Chemical
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Concentrate, 10N ACS (Certified), Fisher Chemical
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution, 0.01N (N/100) (Certified), Fisher Chemical™
Standardized at 25°C to pH 8.2 against sodium hydroxide that has been standardized against NIST™ potassium acid phthalate | CAS: 7647-01-0 | ClH | 36.46 g/mol
Hydrochloric Acid, ACS, Fisher Chemical
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440 PubChem CID: 24290 ChEBI: CHEBI:53434 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution, 0.02N (N/50) (Certified), Fisher Chemical
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid (Technical), Fisher Chemical™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid, ACS Grade, 36.5 to 38.0%, LabChem™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl
Hydrochloric Acid Solution (0.2N (N/5)/Certified), Fisher Chemical™
CAS: 7647-01-0 Molecular Formula: ClH Molecular Weight (g/mol): 36.46 MDL Number: MFCD00011324 MFCD00792839 InChI Key: VEXZGXHMUGYJMC-UHFFFAOYSA-N Synonym: hydrochloric acid,hydrogen chloride,muriatic acid,chlorohydric acid,acide chlorhydrique,chlorwasserstoff,spirits of salt,hydrogen chloride hcl,anhydrous hydrochloric acid,chloorwaterstof PubChem CID: 313 ChEBI: CHEBI:17883 IUPAC Name: hydrogen chloride SMILES: Cl