missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Iodine, 98%, ACS DEA List I, Reagents
Supplier: Labchem Inc C221750025C1
Avertissement:
DEA LIST I
Description
- Iodine, Purified, Lab Grade
- Brand: Reagents
- DEA List I
- Appearance: Violet-black Crystals
- Molecular Formula I2
- MW (g/mol): 253.8089
- Reagents
- Assay >= 98.0%
Spécifications
| Crystals and/or Chunks | |
| 253.81 | |
| Purified |
| 25 g | |
| 98.00% |
Identifiants chimiques
| 253.81 |