missing translation for 'onlineSavingsMsg'
Learn More

Iodine, 98%, ACS DEA List I, Reagents

Catalog No. p-200104732
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
361059162 100 g
361059154 25 g
361059153 2 kg
361059173 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
DEA LIST I
Catalog No. 361059162 Supplier Labchem Inc Supplier No. C2217500100C2
Only null left
DEA LIST I
  • Iodine, Purified, Lab Grade
  • Brand: Reagents
  • DEA List I
  • Appearance: Violet-black Crystals
  • Molecular Formula I2
  • MW (g/mol): 253.8089
  • Reagents
  • Assay >= 98.0%

Chemical Identifiers

Molecular Weight (g/mol) 253.81

Specifications

Physical Form Crystals and/or Chunks
Quantity 100 g
Percent Purity 98.00%
Grade Purified