missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Hydrochloride, 50 g + 100 mL Water, Ricca Chemical

Supplier:  Ricca Chemical Company 392516

 Afficher plus de versions de ce produit

Numéro de catalogue. 3925-16


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Spécifications

Spécifications

Hydroxylamine Hydrochloride
7732-18-5
Laboratory
3.2
Liquid
Colorless
dihydrogen oxide, dihydrogen monoxide
Natural Poly Bottle
500 mL
50g + 100mL Water

Identifiants chimiques

7732-18-5
dihydrogen oxide, dihydrogen monoxide
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS Documents

Documents

Promotions

Promotions