missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrochloric acid concentrate, For 10L standard solution, 1Â M HCl (1N), Honeywell Fluka™
for 10L standard solution, 1Â M HCl (1N)
Supplier: Honeywell-Fluka 382834X1EA
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Spécifications
| Hydrochloric acid concentrate | |
| 7647-01-0,7732-18-5 | |
| ≥70.0000%,≥20.0000% | |
| 1.012 g/cm3 | |
| Poly Bottle | |
| 4 x 1 Ea. | |
| Concentrate | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| Cl | |
| 36.46 | |
| CHEBI:17883 |
| For 10L standard solution, 1Â M HCl (1N) | |
| 100°C (literature) | |
| 1 M HCl (1N) | |
| <80.0000%,<30.0000% | |
| 190 mmHg (25°C) | |
| ClH | |
| HCl | |
| UN1789 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| hydrogen chloride | |
| 313 | |
| 36.46 |
Identifiants chimiques
| 7647-01-0 | |
| 36.46 | |
| VEXZGXHMUGYJMC-UHFFFAOYSA-N | |
| 313 | |
| hydrogen chloride |
| ClH | |
| MFCD00011324 MFCD00792839 | |
| hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof | |
| CHEBI:17883 | |
| Cl |
Sécurité et manipulation
P305 + P351 + P338
missing translation for 'shelfLife' : 1260 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Hydrochloric acid
Recommended Storage : Room Temp