missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric acid concentrate, For 10L standard solution, 1 M HCl (1N), Honeywell Fluka™

for 10L standard solution, 1 M HCl (1N)

$1974.35 - $1974.35

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
60046715
View Documents
Honeywell Inc
382834X1EA
4 x 1 Ea.
Each for $1,974.35
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7647-01-0
36.46
VEXZGXHMUGYJMC-UHFFFAOYSA-N
313
hydrogen chloride
ClH
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
CHEBI:17883
Cl

Specifications

7647-01-0,7732-18-5
≥70.0000%,≥20.0000%
1.012 g/cm3
Poly Bottle
4 x 1 Ea.
Concentrate
MFCD00011324 MFCD00792839
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Cl
36.46
CHEBI:17883
Hydrochloric acid concentrate, For 10L standard solution, 1 M HCl (1N)
100°C (literature)
1 M HCl (1N)
<80.0000%,<30.0000%
190 mmHg (25°C)
ClH
HCl
UN1789
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrogen chloride
313
36.46
Videos
Safety and Handling

Safety and Handling

P305 + P351 + P338

ShelfLife : 1260 days from date of manufacture

DOTInformation : Transport Hazard Class: 8; Packing Group:3: II; Proper Shipping Name: Hydrochloric acid

Recommended Storage : Room Temp

SDS
missing translation for 'documents'

missing translation for 'documents'