Learn More
Cytiva Life Sciences™ HyClone™ Dulbecco's Modified Eagle Medium with Low Glucose: Powder
Catalog No. SH3004403
Change view
Click to view available options
Quantity:
10 L
2 x 5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3004403 10 L
SH3004402 2 x 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3004403 Supplier Cytiva Life Sciences™ Supplier No. SH30044.03
Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions
Supports cell growth in research and biomanufacturing cell culture processes.
Supports cell growth in research and biomanufacturing cell culture processes.
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
  • Raw material components screened through strict quality control testing for lot-to-lot consistency.
  • Products are hydrated using purified process water and undergo sterile filtration.
Specifications
Form Powder
Product Type Modified Eagle Medium
Quantity 10 L
Description Low Glucose