missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human TAF15 (aa 117-184) Control Fragment Recombinant Protein
Recombinant Protein
Supplier: Invitrogen™ RP101497
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62543 (PA5-62543. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Initiation of transcription by RNA polymerase II requires the activities of more than 70 polypeptides. The protein that coordinates these activities is transcription factor IID (TFIID), which binds to the core promoter to position the polymerase properly, serves as the scaffold for assembly of the remainder of the transcription complex, and acts as a channel for regulatory signals.Spécifications
| Q92804 | |
| Blocking Assay, Control | |
| 8148 | |
| 100 μL | |
| 2610111C21Rik; 68 kDa TATA-binding protein-associated factor; 68 kDa; hTAFII68; LOC100713096; Npl3; RBP56; RBP56/CSMF fusion; RNA-binding protein 56; TAF(II)68; Taf15; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68 kDa; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68 kDa; TAF2N; TAFII68; TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68 kD (RNA binding protein 56); TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68 kD (RNA-binding protein 56); TATA box binding protein associated factor 15; TATA box-binding protein-associated factor 2 N (RNA-binding protein 56); TATA-bin; TATA-binding protein-associated factor 2 N; TATA-box binding protein associated factor 15; TBP-associated factor 15 | |
| TAF15 | |
| Human | |
| His-ABP-tag | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ≥5.0 mg/mL | |
| 1 M urea, PBS with no preservative; pH 7.4 | |
| Human TAF15 (aa 117-184) Control Fragment | |
| RUO | |
| TAF15 | |
| Unconjugated | |
| Recombinant | |
| DQQSGYDQHQGSYDEQSNYDQQHDSYSQNQQSYHSQRENYSHHTQDDRRDVSRYGEDNRGYGGSQGGG | |
| E. coli | |
| >80% by SDS-PAGE and Coomassie blue staining |