Learn More
Invitrogen™ Human PDGF-BB ELISA Kit
ELISA
$856.00 - $5960.00
Spécifications
| Accession Number | P01127 |
|---|---|
| Assay Range | 31.3-2000 pg/mL |
| Assay Sensitivity | 4.6 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
Comprend: Aluminium pouch(es) with a Microwell Plate coated with antibody to human PDGF-BB
Biotin-Conjugate anti-human PDGF-BB antibody
Streptavidin-HRP
Human PDGF-BB Standard lyophilized, 4ng/mL upon reconstitution
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
50246659
|
Invitrogen™
BMS2071 |
96 Tests |
chaque for $856.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
50246660
|
Invitrogen™
BMS2071TEN |
10 x 96 Tests |
chaque for $5,960.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
The Human Platelet Derived Growth Factor BB (Hu PDGF-BB) ELISA quantitates Hu PDGF-BB in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu PDGF-BB. Principle of the method The Human PDGF-BB solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Spécifications
| P01127 | |
| 4.6 pg/mL | |
| ELISA | |
| Human | |
| Absorbance Microplate Reader | |
| PDGFB | |
| 4% | |
| Biotin | |
| Plasma 20 μL, Serum 20 μL, Supernatant 20 μL | |
| Platelet derived growth factor BB | |
| 1 hr 20 min |
| 31.3-2000 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| ELISA | |
| 5155 | |
| 8.4% | |
| Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2-8°C | |
| PDGF-BB | |
| 3 hr 30 min |