missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human IL-26 ELISA Kit
ELISA
$874.00 - $4330.00
Spécifications
| Accession Number | AK155; il 26; IL26; IL-26; ILN; Interleukin; interleukin 26; interleukin-26; Protein AK155 |
|---|---|
| Assay Range | 3.3 to 800 pg/mL |
| Assay Sensitivity | 3.3 pg/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
EH270RBX5
|
Invitrogen™
EH270RBX5 |
5 x 96 Tests |
chaque for $4,330.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
EH270RB
|
Invitrogen™
EH270RB |
96 Tests |
chaque for $874.00
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Human IL-26 quantitates human IL-26 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human IL-26.
IL26 is a member of the IL10 family of cytokines. It is a secreted protein and may function as a homodimer. This protein is thought to contribute to the transformed phenotype of T cells after infection by herpesvirus samimiri.Spécifications
| AK155; il 26; IL26; IL-26; ILN; Interleukin; interleukin 26; interleukin-26; Protein AK155 | |
| 3.3 pg/mL | |
| ELISA | |
| Colorimetric Microplate Reader | |
| IL26 | |
| <10 | |
| HRP | |
| Plasma 50 μL, Serum 50 μL, Supernatant 50 μL | |
| IL-26 | |
| 4 hrs 45 mins |
| 3.3 to 800 pg/mL | |
| HRP | |
| Plasma, Serum, Supernatant | |
| 55801 | |
| <12% | |
| Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
| RUO | |
| 2°C to 8°C | |
| 1 hrs 20 mins |