missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FGFBP3 (aa 32-63) Control Fragment Recombinant Protein

Recombinant Protein

Supplier:  Invitrogen™ RP101501

Numéro de catalogue. RP101501

Il en reste null
Découvrez plus d'offres spéciales

Description

Description

Highest antigen sequence indentity to the following orthologs: Mouse (59%), Rat (59%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-62879 (PA5-62879. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

FGFBP3 is a heparin-binding protein which binds to FGF2, prevents binding of FGF2 to heparin and probably inhibits immobilization of FGF2 on extracellular matrix glycosaminoglycans, allowing its release and subsequent activation of FGFR signaling which leads to increased vascular permeability.
Spécifications

Spécifications

Q8TAT2
Blocking Assay, Control
143282
100 μL
2610306H15Rik; C10orf13; FGF-binding protein 3; Fgfbp3; Fgf-bp3; FGFBP-3; fibroblast growth factor binding protein 3; fibroblast growth factor-binding protein 3; PSEC0101
FGFBP3
Human
His-ABP-tag
-20°C, Avoid Freeze/Thaw Cycles
Liquid
≥5.0 mg/mL
1 M urea, PBS with no preservative; pH 7.4
Human FGFBP3 (aa 32-63) Control Fragment
RUO
FGFBP3
Unconjugated
Recombinant
AASNVAEPVPGPTGGSSGRFLSPEQHACSWQL
E. coli
>80% by SDS-PAGE and Coomassie blue staining
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions