Learn More
Gibco™ Human CCL18 (MIP-4) Recombinant Protein
Recombinant Protein
Supplier: Gibco™ PHC1254
Description
Carrier-Free Reconstitution: We recommend that this vial be briefly centrifuged prior to opening to bring the contents to the bottom. Reconstitute MIP-4 in sterile, distilled water to a concentration of 0.1-1.0 mg/mL. Do not vortex. This solution can be stored at 2°C to 8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein, such as 0.1% BSA and store in working aliquots at -20°C to -80°C. Storage: The lyophilized preparation is stable at room temperature for up to 1 month, but should be stored at -20°C for long term storage. Working aliquots stored with a carrier protein are stable for at least 3 months at -20°C to -80°C. Avoid repeated freeze/thaw cycles.
Macrophage Inflammatory Protein-4 (MIP-4), also called CCL18, is a chemokine expressed in the lymph nodes, lungs, placenta and bone marrow. Although the MIP-4s receptor remains undetermined, MIP-4 is known to act as a chemoattractant for activated and non-activated T cells.Spécifications
| P55774 | |
| Protein with no preservative | |
| 7.8 kDa | |
| Purified | |
| -20°C | |
| <0.1 ng/μg | |
| PARC | |
| Protein | |
| Human | |
| Human CCL18 recombinant protein contains 68 amino acids | |
| Carrier-Free | |
| Wet Ice | |
| Chemokines & Receptors | |
| Lyophilized | |
| Immunology, Inflammation, Signal Transduction, Neurobiology | |
| >98% by SDS-PAGE |
| Bioactivity | |
| 6362 | |
| Human CCL18 (MIP-4) | |
| 10 μg | |
| RUO | |
| Alternative macrophage activation-associated CC chemokine 1; AMAC1; AMAC-1; CC chemokine ligand 18; CC chemokine PARC; C-C motif chemokine 18; C-C motif chemokine ligand 18; CCL18; CCL18(1-68); CCL18(3-69); CCL18(4-69); chemokine (C-C motif) ligand 18 (pulmonary and activation-regulated); chemokine (C-C), dendritic; CKb7; CUL9; CUL-9; cullin 9; cullin-9; DCCK1; DC-CK1; dendritic cell chemokine 1; H7AP1; H-MIP-4; KIAA0708; Macrophage inflammatory protein 4; MIP4; MIP-4; p53-associated parkin-like cytoplasmic protein; PARC; parkin-like cytoplasmic p53 binding protein; pulmonary and activation-regulated chemokine; SCYA18; small inducible cytokine A18; small inducible cytokine subfamily A (Cys-Cys), member 18, pulmonary and activation-regulated; small-inducible cytokine A18; UbcH7-associated protein 1 | |
| CCL18 | |
| Unconjugated | |
| Recombinant | |
| -20°C | |
| <0.1 ng/μg | |
| E. coli | |
| Recombinant, Ligand | |
| MIP (Macrophage Inflammatory Protein) Chemokines | |
| Gibco |