Learn More
Azure Biosystems HRP Stripping Buffer
Stripping buffer uniformly removes antibodies from HRP Western blot before reprobing with another set of antibodies.
Supplier:  Azure Biosystems AC2154
Numéro de catalogue. AC2154AZ
Il en reste null
Description
Description
  • Works by removing the primary and secondary antibodies from the membrane
  • Stripping buffer for stripping and reprobing chemiluminescent Western blots. Provided as a 1x ready-to-use solution
  • Size: 500 mL
Spécifications
Spécifications
Western Blot Stripping Buffer
Liquid
Western Blot
500 mL
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS