Learn More
Invitrogen™ HIF-2 alpha Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA116510
Numéro de catalogue. APPA6510
Il en reste null
Découvrez plus d'offres spéciales
Description
Description
Suggested positive control: Cos7 CoCl2-treated nuclear extract, PC12 nuclear extracts.
HIF-2 alpha (Epas1) is a transcription factor involved in the induction of genes regulated by oxygen. HIF-2 alpha contains a basic-helix-loop-helix domain protein dimerization domain as well as a domain found in proteins in signal transduction pathways which respond to oxygen levels. It also regulates the vascular endothelial growth factor (VEGF) expression and seems to be implicated in the development of blood vessels and the tubular system of the lungs. Mutations in this gene are associated with erythrocytosis familial type 4.
Spécifications
Spécifications
HIF-2 alpha
Polyclonal
Unconjugated
EPAS1
basic-helix-loop-helix-PAS protein MOP2; bHLHe73; Class E basic helix-loop-helix protein 73; ECYT4; endothelial PAS domain protein 1; endothelial PAS domain protein 1b; endothelial PAS domain-containing protein 1; endothelial PAS domain-containing protein 1b; EPAS; Epas1; EPAS-1; epas1b; Hif like protein; HIF1 alpha-like factor; HIF-1-alpha-like factor; HIF-1alpha-like factor; HIF1alpha-like factor; HIF2 alpha; HIF-2 alpha; HIF2A; hif-2a; hif-2ab; HIF-2alpha; HIF-2-alpha; HIF2-alpha; HIF-related factor; HLF; HLF (HIF1alpha-like factor); HRF; hypoxia inducible factor 2 alpha; hypoxia inducible factor 2, alpha subunit; hypoxia inducible factor 2a; hypoxia inducible transcription factor 2alpha; hypoxia-inducible factor 2 alpha; hypoxia-inducible factor 2-alpha; Member of PAS protein 2; mHLF; MOP2; PAS domain-containing protein 2; PASD2; wu:fq36f01
Rabbit
Antigen affinity chromatography
RUO
100348809, 100771160, 13819, 2034, 29452, 443286
-20°C
Liquid
ChIP Assay, ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.05% sodium azide
P97481, Q99814, Q9JHS1
EPAS1
A peptide derived from the C-terminus of mouse/human HIF-2 alpha protein.
100 μL
Primary
Fish, Hamster, Human, Mouse, Monkey, Sheep, Rabbit, Reptile, Rat
Antibody
IgG
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS