missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexadecylbenzene 99.0+%, TCI America™
Supplier: TCI America H05135G
Spécifications
| Hexadecylbenzene | |
| White | |
| C22H38 | |
| 5 g | |
| DEQLTFPCJRGSHW-UHFFFAOYSA-N | |
| hexadecylbenzene | |
| 15085 | |
| ≥99.0% (GC) |
| 1459-09-2 | |
| 172°C | |
| MFCD00039871 | |
| 1-phenylhexadecane, benzene, hexadecyl, n-hexadecylbenzene, hexadecane, 1-phenyl, phenyl hexadecane, unii-39hl93ne0r, cetylbenzene, acmc-1bt4a, hexadecane, 1 or 16-phenyl | |
| CCCCCCCCCCCCCCCCC1=CC=CC=C1 | |
| 302.55 | |
| 302.55 | |
| Crystalline Lumps |
Identifiants chimiques
| 1459-09-2 | |
| 302.55 | |
| DEQLTFPCJRGSHW-UHFFFAOYSA-N | |
| 15085 | |
| CCCCCCCCCCCCCCCCC1=CC=CC=C1 |
| C22H38 | |
| MFCD00039871 | |
| 1-phenylhexadecane, benzene, hexadecyl, n-hexadecylbenzene, hexadecane, 1-phenyl, phenyl hexadecane, unii-39hl93ne0r, cetylbenzene, acmc-1bt4a, hexadecane, 1 or 16-phenyl | |
| hexadecylbenzene |
Sécurité et manipulation
missing translation for 'einecsNumber' : (3)-0021
missing translation for 'tsca' : Yes