missing translation for 'onlineSavingsMsg'
Learn More

Hexadecylbenzene 99.0+%, TCI America™

Catalog No. p-7130575
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H05135G 5 g
H051325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H05135G Supplier TCI America Supplier No. H05135G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1459-09-2
Molecular Formula C22H38
Molecular Weight (g/mol) 302.55
MDL Number MFCD00039871
InChI Key DEQLTFPCJRGSHW-UHFFFAOYSA-N
Synonym 1-phenylhexadecane, benzene, hexadecyl, n-hexadecylbenzene, hexadecane, 1-phenyl, phenyl hexadecane, unii-39hl93ne0r, cetylbenzene, acmc-1bt4a, hexadecane, 1 or 16-phenyl
PubChem CID 15085
IUPAC Name hexadecylbenzene
SMILES CCCCCCCCCCCCCCCCC1=CC=CC=C1

Specifications

Color White
Boiling Point 172°C
Quantity 5 g
Formula Weight 302.55
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material Hexadecylbenzene
EINECSNumber (3)-0021
TSCA Yes