missing translation for 'onlineSavingsMsg'
Learn More

Hexachloroethane, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Supplier:  SPEX CertiPrep Group LLC S2155

Numéro de catalogue. S2155


Il en reste null
Découvrez plus d'offres spéciales

Comprend: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Hexachloroethane67-72-11000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Spécifications

Spécifications

Hexachloroethane
67-72-1,67-56-1
Certificate of Analysis reports target analytes to a ±2% accuracy and correct for raw material purity.
C2Cl6
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
VHHHONWQHHHLTI-UHFFFAOYSA-N
Methanol-P and T
236.72
6214
2mL amber prescored glass ampule
0.1,99.9
0.1,99.9
MFCD00000799
Certified Reference Material
ClC(Cl)(Cl)C(Cl)(Cl)Cl
hexachloroethane
1 mL
CHEBI:39227

Identifiants chimiques

67-72-1
236.72
VHHHONWQHHHLTI-UHFFFAOYSA-N
CHEBI:39227
ClC(Cl)(Cl)C(Cl)(Cl)Cl
C2Cl6
MFCD00000799
6214
hexachloroethane
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'dotInformation' : Nonhazardous

Recommended Storage : Ambient

FDS
Documents

Documents

Promotions

Promotions

Product intended for laboratory use only