missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ HDPE, Carboy with Spigot

For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Closure Material: Polypropylene; Color: White

$363.55 - $382.16

Spécifications

Closure Material Polypropylene
Color White
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders.
Material HDPE
Product Type Carboy with Spigot
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Prix Quantité  
19-830-415
Afficher les documents
Justrite™
JMC12914
5 L
chaque for $363.55
Il en reste null
 
19-830-416
Afficher les documents
Justrite™
JMC12915
10 L
chaque for $382.16
Il en reste null
 
Description

Description

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Material, volume, and metric graduation marks imprinted on carboys
Spécifications

Spécifications

Polypropylene
For storing and dispensing solvents, buffers, reagents and powders.
Carboy with Spigot
White
HDPE
Rectangular
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'