Learn More
Eisco™ Class B Glass Graduated Cylinder with Plastic Base and Guard
Graduated Borosilicate glass graduated cylinder with a plastic guard and detachable Hexagonal base.
$9.97 - $72.50
Spécifications
Certifications/Compliance DIN EN ISO 4788
Class Class B
Scale Color Blue
Shape Round
Sterility Non-sterile
voir les spécifications
Produits 6
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Graduations Type Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Capacity (Metric) Graduations Type Prix Quantité  
S27968
Afficher les documents
Eisco™
CH0346A
10 mL 0.2 mL Graduated Measuring Cylinder
chaque for $9.97
Il en reste null
S15492
Afficher les documents
Eisco™
CH0346SET
10 mL, 50 mL, 100 mL 1 mL Graduated Cylinder Set
chaque for $72.50
Il en reste null
S88457
Afficher les documents
Eisco™
CH0346B
25 mL 0.5 mL Graduated Measuring Cylinder
chaque for $15.10
Il en reste null
S27970
Afficher les documents
Eisco™
CH0346D
50 mL 1 mL Graduated Measuring Cylinder
chaque for $12.50
Il en reste null
S27969
Afficher les documents
Eisco™
CH0346C
100 mL 1 mL Graduated Measuring Cylinder
chaque for $13.87
Il en reste null
S27971
Afficher les documents
Eisco™
CH0346E
250 mL 2 mL Graduated Measuring Cylinder
chaque for $24.50
Il en reste null
Description
Description
  • Made from high quality Borosilicate 3.3 Glass
  • Detachable, plastic hexagonal base and protective collar
  • Hexagonal base for maximum stability
  • Plastic base and plastic gaurd are removable for easy cleaning
  • Printed graduations are chemical resistant
  • Spout for easy, mess free pouring
  • Class B
  • Conforms to DIN EN ISO 4788 standard
  • Autoclavable
Spécifications
Spécifications
DIN EN ISO 4788
Blue
Non-sterile
Measuring Liquids
Class B
Round
Borosilicate Glass
Autoclavable
Videos
FDS
missing translation for 'documents'
missing translation for 'documents'