Learn More
Corning™ Petri Dishes
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
$424.21 - $1,044.53
| Catalog Number | Mfr. No. | Dimensions (Dia. x H) | Well Count | Vent | Sterility | Price | Quantity & Availability | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (Dia. x H) | Well Count | Vent | Sterility | Price | Quantity & Availability | ||
07201913
![]() |
Corning™
BB6401 |
65 x 14.5 mm | 1 | 6 | Sterile |
Case of 720 Each for $1,044.53
|
|
||
07202013
![]() |
Corning™
BP6310 |
65 x 14.5 mm | 1 | 3 | Sterile |
Case of 720 Each for $519.33
|
|
||
07202027
![]() |
Corning™
BP93B101 |
90 x 14.2 mm | 1 | 3 | Aseptically Manufactured |
Case of 825 Each for $424.21
|
|
||
07202028
![]() |
Corning™
BP93B102 |
100 x 15 mm | 1 | 3 | Sterile |
Case of 825 Each for $465.21
|
|
||
07202029
![]() |
Corning™
BP93D102 |
90 x 14.2 mm | 1 | 3 | Sterile |
Case of 825 Each for $464.88
|
|
||
07202050
![]() |
Corning™
BP5303 |
55 x 14.2 mm | 1 | 6 | Sterile |
Case of 1620 Each for $952.61
|
|
||
07202550
![]() |
Corning™
BP14306 |
140 x 20.6 mm | 1 | 6 | Sterile |
Case of 176 Each for $451.53
|
|
||
07201995
![]() |
Corning™
BP12405 |
120.5 x 15.8 mm | 1 | 4 | Sterile |
Case of 252 Each for $1,040.75
|
|
||
Description
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
- Crystal polystyrene
- Optimal flatness
- Manufactured in class 100
- Easy opening