missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Petri Dishes

Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
Supplier: Corning™ BP14306
Description
Optimized for automation. Gosselin™ Petri Dishes are manufactured from crystal polystyrene.
Sterile product features a double outer bag with traceability. Optimized for automation.- Three vents
- Crystal polystyrene
- Easy opening sleeves
- Sterile
Specifications
| Clear | |
| Round | |
| 1 | |
| 6 | |
| 20.6 mm |
| Polystyrene | |
| Sterile | |
| Optimized for automation | |
| 140 x 20.6 mm | |
| 140 mm |