missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Compass Biomedical™ GMP PLUS™ Cryobag Human Platelet Lysate
Advanced human platelet lysate that is safe and consistent for the rapid expansion of human mesenchymal cell cultures.
Spécifications
| For Use With (Application) | Cell Culture, Stem Cell Culture |
|---|---|
| Purity | GMP Grade |
| Host Species | Human |
| Product Type | Human Platelet Lysate |
| Content And Storage | Store at -80°C upon receipt. Thaw at room temperature or in a water bath. After thawing, aliquot and store at -20°C or colder. Long term storage at -80°C is recommended. Multiple freeze/thaw cycles are not recommended. |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantité | |||||
|
PLS5
|
Compass Biomedical™
PLS5 |
100 mL |
N/A
|
N/A | |||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
|
PLS6
|
Compass Biomedical™
PLS6 |
250 mL |
N/A
|
N/A | |||||
| Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437. | |||||||||
Description
- Potent: Human derived product rich in growth factors, cytokine, and other proteins
- Products are growth factor rich supplements made from large pools of transfusion grade human platelets sourced from FDA-registered/AABB accredited blood centers.
Spécifications
| Cell Culture, Stem Cell Culture | |
| Human | |
| Store at -80°C upon receipt. Thaw at room temperature or in a water bath. After thawing, aliquot and store at -20°C or colder. Long term storage at -80°C is recommended. Multiple freeze/thaw cycles are not recommended. | |
| GMP |
| GMP Grade | |
| Human Platelet Lysate | |
| Cryobag |